Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
9-Fluorenylmethyl carbamate
9-Fluorenylmethyl carbamate 99%
9-Fluorenylmethyl carbamate 99%
:
Thermo Fisher Scientific
9-Fluorenylmethyl carbamate 99%
:
Thermo Fisher Scientific
:
84418-43-9
:
:
C₁₅H₁₃NO₂
:
MFCD00237376
:
239.27 g/mol
:
204…207 °C (dec.)
:
GHS07
:
Ambient
Appearance:
White to off-white Powder
Infrared spectrum:
Conforms
HPLC:
≥98.5 %
ACRO43417