Order Entry
Germany
ContactUsLinkComponent
Toluene-[D₈] 99.5 atom % D, AcroSeal® for NMR spectroscopy
Flammable
Irritant
Health
Toluene-[D₈] 99.5 atom % D, AcroSeal® for NMR spectroscopy
Catalog # 434001000.
CAS Number:  
undefined
Flammable
Irritant
Health
Toluene-[D₈] 99.5 atom % D, AcroSeal® for NMR spectroscopy
Catalog # 434001000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤50 ppm
  • D-Enrichment:
    ≥99.4 %
  • GC:
    ≥96.0 %
  • NMR (1H-NMR):
    Consistent with structure

Specification Test Results

About this item

 :
 :100.08 g/mol
Boiling Pt:110 °C (760 mmHg)
Melting Pt:–84 °C
Density:0.94 g/cm³ (20 °C)
Flash Pt:4 °C
UN:1294
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:2037-26-5
Formula:C₆D₅CD₃
MDL Number:MFCD00044638