
N'-Hydroxypyridine-4-carboximidamide 97%
Catalog # ACRO432480050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to light yellow Crystalline solid
- Infrared spectrum:Conforms
- Melting point:200 to 207 °C
- HPLC:≥96.0 %
Specification Test Results
About this item
:
:137.142 g/mol
Melting Pt:175…180 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1594-57-6
Formula:C₆H₇N₃O
MDL Number:MFCD00125873