Order Entry
Germany
ContactUsLinkComponent
4,5,6,7-Tetrahydropyrazolo[1,5-a]pyridine-3-carbaldehyde 97%
Irritant
4,5,6,7-Tetrahydropyrazolo[1,5-a]pyridine-3-carbaldehyde 97%
Catalog # ACRO431090010
CAS Number:  
undefined
Irritant
4,5,6,7-Tetrahydropyrazolo[1,5-a]pyridine-3-carbaldehyde 97%
Catalog # ACRO431090010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to yellow to orange Low melting solid or liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :150.181 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:307308-03-8
Formula:C₈H₁₀N₂O
MDL Number:MFCD09834817