Order Entry
Germany
ContactUsLinkComponent
 
 
3-Iodo-4-pyridine carboxylic acid 97%
 
undefined
 
3-Iodo-4-pyridine carboxylic acid 97%

 

  • Appearance:
    Off-white to beige Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    240 to 244 °C
  • HPLC:
    ≥96.0 %