
6-Quinolineboronic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:376581-24-7
:
Formula:C₉H₈BNO₂
MDL Number:MFCD03093100
:172.98 g/mol
Melting Pt:144…145 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
- GC:≥99.0 %
- Color scale:≤25 APHA
- Stabilizer:Approx. 50 ppm BHT
- Non-volatile matter:≤0.005 %
- Acidity:≤0.0004 meq/g
- Aluminium (Al):≤0.5 ppm
- Barium (Ba):≤0.1 ppm
- Calcium (Ca):≤0.5 ppm
- Cobalt (Co):≤0.02 ppm
- Copper (Cu):≤0.02 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.1 ppm
- Magnesium (Mg):≤0.1 ppm
- Manganese (Mn):≤0.02 ppm
- Nickel (Ni):≤0.02 ppm
- Tin (Sn):≤0.1 ppm
- Zinc (Zn):≤0.1 ppm
- Peroxides:≤0.005 % (as H2O2)