Order Entry
Germany
ContactUsLinkComponent
 
 
1-Methyl-1H-indazole-6-boronic acid 97%
  ACRO430480010
 :  
undefined
 
1-Methyl-1H-indazole-6-boronic acid 97%
  ACRO430480010
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥96.0 %
  • Enantiomeric excess:
    ≥98.0 %

 

 

 :
 :175.9803 g/mol
 :259.8…261.2 °C
 :GHS07
 :Ambient
 :1150114-80-9
 :C₈H₉BN₂O₂
 :MFCD09870053