Order Entry
Germany
ContactUsLinkComponent
Hydrogen chloride ∼0.5 M in methanol, AcroSeal®
Flammable
Toxic
Health
Hydrogen chloride ∼0.5 M in methanol, AcroSeal®
Catalog # 428931000.
CAS Number:  
undefined
Flammable
Toxic
Health
Hydrogen chloride ∼0.5 M in methanol, AcroSeal®
Catalog # 428931000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Solution
  • Strength:
    0.4 to 0.6 M
  • Melting point:
    143 to 149 °C
  • Titration Argentometric:
    ≥94 %

Specification Test Results

About this item

 :
 :36.46 g/mol
Density:0.8 g/cm³
Flash Pt:12 °C
Storage Temperature:Refrigerator
UN:3286
ADR:3,II
 :GHS02
 :GHS06
 :GHS08
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324