Order Entry
Germany
ContactUsLinkComponent
 
Di-tert-butyl-N,N-diisopropylphosphoramidite 95%
 
undefined
Di-tert-butyl-N,N-diisopropylphosphoramidite 95%

 

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4390 to 1.4410 (20°C, 589 nm)
  • GC:
    ≥94 %