



1-Propynylmagnesium bromide 0.5 M in THF, AcroSeal®
:16466-97-0
:
:C₃H₃BrMg
:MFCD00216588
:143.27 g/mol
:65…67 °C (760 mmHg)
:0.94 g/cm³
:–1 °C
:3399
:4.3,I
:GHS07
:GHS05
:GHS02
:GHS08
:Ambient
- Appearance:Clear colorless to yellow Solution (May develop some turbidity or precipitate)
- Titration Grignard:0.45 to 0.65 M
- Refractive index:1.5270 to 1.5310 (20°C, 589 nm)
- GC:≥95 %