Order Entry
Germany
ContactUsLinkComponent
2-Fluoro-m-anisaldehyde 96%
 
2-Fluoro-m-anisaldehyde 96%
  ACRO425830010
 :  
 
2-Fluoro-m-anisaldehyde 96%
  ACRO425830010
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to yellow Crystalline solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    44 to 48 °C
  • GC:
    ≥95 %

 

 

 :
 :154.14 g/mol
 :47…51 °C
 :>110 °C
 :GHS07
 :Ambient
 :103438-88-6
 :C₈H₇FO₂
 :MFCD03411962