tert-Butyl pyrrolidine-1-carboxylate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:86953-79-9
:
Formula:C₉H₁₇NO₂
MDL Number:MFCD00216581
:171.24 g/mol
Boiling Pt:80 °C (.2 mmHg)
Density:0.977 g/cm³
Flash Pt:86 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
- NMR (1H-NMR):Consistent with structure