Methyl-4,4-dimethyl-3-oxovalerate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:55107-14-7
:
Formula:C₈H₁₄O₃
MDL Number:MFCD00008847
:158.20 g/mol
Boiling Pt:67…70 °C (13 mmHg)
Density:0.98 g/cm³
Flash Pt:78 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- GC:≥97.5 %
- HPLC:≥97.5 %