Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Dichloro-p-benzoquinone
2,5-Dichloro-p-benzoquinone 98%
2,5-Dichloro-p-benzoquinone 98%
:
Thermo Fisher Scientific
2,5-Dichloro-p-benzoquinone 98%
:
Thermo Fisher Scientific
:
615-93-0
:
:
C₆H₂Cl₂O₂
:
MFCD00041736
:
159…161 °C
:
176.99 g/mol
:
GHS07
:
Ambient
Appearance:
Yellow to orange Crystalline powder
Infrared spectrum:
Conforms
Melting point:
159 to 164 °C
GC:
≥97.5 %
ACRO40673