L(+)-allo-Isoleucine 99%, 99% ee
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1509-34-8
:
Formula:H₃CCH₂CH(CH₃)CH(NH₂)COOH
MDL Number:MFCD00066446
:131.17 g/mol
Melting Pt:285 °C
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- NMR:consistent with structure
- Enantiomeric excess:≥98.5 % (HPLC)
- Specific optical rotation:+35° to +39° (20°C, 589 nm) (C=5, 1N HCl)