
9,9-Di-n-octylfluorene-2,7-diboronic acid 98%
ACRO398310050
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
:
:478.287 g/mol
:158…162 °C
:GHS07
:Ambient
:258865-48-4
:C₂₉H₄₄B₂O₄
:MFCD03701608