Order Entry
Germany
ContactUsLinkComponent
 
2-Chloro-5-pyridineboronic acid 97%
 
undefined
2-Chloro-5-pyridineboronic acid 97%

 

  • Appearance:
    Yellow to orange Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    161 to 167 °C
  • HPLC:
    ≥96.0 %