7-Methoxyindole 97%
Catalog # ACRO397520010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to brown Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
Specification Test Results
About this item
:
:147.18 g/mol
Boiling Pt:166 °C (27 mmHg)
Density:1.126 g/cm³
Flash Pt:113 °C
:GHS07
Storage Temperature:Ambient
CAS Number:3189-22-8
Formula:C₉H₉NO
MDL Number:MFCD00047203