Order Entry
Germany
ContactUsLinkComponent
(4-Boc-aminophenyl)boronic acid 97%
(4-Boc-aminophenyl)boronic acid 97%
Catalog # ACRO397030010
CAS Number:  
undefined
(4-Boc-aminophenyl)boronic acid 97%
Catalog # ACRO397030010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to brown Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :237.062 g/mol
Melting Pt:199…204 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:380430-49-9
Formula:C₁₁H₁₆BNO₄
MDL Number:MFCD02093054