Nickel activated 50% slurry in water catalyst (Raney®-type)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-02-0
:
Formula:Ni
MDL Number:MFCD00011137
:58.69 g/mol
Boiling Pt:3007 °C (1013 hPa)
Melting Pt:1453 °C
Density:8,9 g/cm³ (20 °C)
UN:1378
ADR:4.2,II
:GHS07
:GHS08
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark grey to black Slurry in water
- pH:8.5 to 12.0 (of supernatant)
- Aluminium (Al):≤8 %
- Nickel (Ni):≥92 %