Order Entry
Germany
ContactUsLinkComponent
Charcoal activated, NORIT® GAC 1240 12-40 mesh
Charcoal activated, NORIT® GAC 1240 12-40 mesh
Certificates
undefined
Charcoal activated, NORIT® GAC 1240 12-40 mesh
Certificates

Specification Test Results

  • Appearance:
    Black Granular solid
  • Water:
    ≤2 % (As packed)
  • Particle size:
    ≤5 % (> 12 mesh)
  • Molasses number:
    ≥230
  • Abrasion number:
    ≥75
  • Iodine number:
    ≥1000 mg/g

Specification Test Results

Related Information