
2,4,5-Trifluorophenylboronic acid 97%
Catalog # ACRO393800050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
Specification Test Results
About this item
:
:175.9 g/mol
Melting Pt:118…119 °C
:GHS07
Storage Temperature:Ambient
CAS Number:247564-72-3
Formula:C₆H₄BF₃O₂
MDL Number:MFCD01863165