4-Iodobenzaldehyde 97+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15164-44-0
:
Formula:C₇H₅IO
MDL Number:MFCD00039576
:232.02 g/mol
Melting Pt:77…78 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to yellow to beige Powder or crystals
- Infrared spectrum:Conforms
- Melting point:75 to 80 °C
- HPLC:≥97.0 %