3-Butynyl toluene-4-sulphonate 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:23418-85-1
:
Formula:C₁₁H₁₂O₃S
MDL Number:MFCD00041687
:224.28 g/mol
Density:1.012 g/cm³
Flash Pt:110 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5215 to 1.5255 (20°C, 589 nm)
- GC:≥95 %