
Tin(IV) iodide 99+% -10 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7790-47-8
:
Formula:I₄Sn
MDL Number:MFCD00036272
:626.33 g/mol
Melting Pt:144 °C
Density:4,46 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3260
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Brown Powder
- Titration Argentometric:≥99.0 %