5'-Bromo-2'-hydroxyacetophenone 98%
Glass bottle, 10 g
Supplier: Thermo Fisher Scientific
5'-Bromo-2'-hydroxyacetophenone 98%
Catalog # ACRO367210100
Supplier: Thermo Fisher Scientific
CAS Number: 1450-75-5
Jump to:
Product Details & Documents
Synonyms: 1-(5-Bromo-2-hydroxyphenyl)ethan-1-one, 1-(5-Bromo-2-hydroxyphenyl)ethanone
- CAS Number:1450-75-5
- Formula:C₈H₇BrO₂
- MDL Number:MFCD00191850
- Molecular Weight:215.05
- Melting Pt:58…61 °C
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:White to yellow to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:57 to 61 °C
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle
- Pk:10 g



