Order Entry
Germany
ContactUsLinkComponent
 
 
 
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) 97%
 
undefined
 
 
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) 97%

 

  • Appearance:
    White to beige Crystalline powder and/or lumps
  • Infrared spectrum:
    Conforms
  • Melting point:
    183 to 193 °C (Dec)
  • HPLC:
    ≥96.0 %