tert-Butylmagnesium chloride 1.7 M in THF, AcroSeal®
:677-22-5
:
:C₄H₉ClMg
:MFCD00000465
:116.87 g/mol
:0.931 g/cm³
:–17 °C
:3399
:4.3,I
:GHS05
:GHS02
:GHS07
:GHS08
:Ambient
- Appearance:Dark brown Solution
- Titration Active base:19 to 21 %