
L-Cysteine methyl ester hydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18598-63-5
:
Formula:C₄H₉NO₂S·HCl
MDL Number:MFCD00038998
:171.7 g/mol
Melting Pt:143…144 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Titration Iodometric:≥97.5 %
- Specific optical rotation:-1.5° to -3.2° (20°C, 589 nm) (c=5, 1 N HCl)
Specification Test Results
Frequently Bought Together