Acetic acid 99+%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:64-19-7
:
Formula:H₃CCOOH
MDL Number:MFCD00036152
:60.05 g/mol
Boiling Pt:117…118 °C (760 mmHg)
Melting Pt:16…16.5 °C
Density:1.048 g/cm³
Flash Pt:40 °C
Storage Temperature:Ambient
UN:2789
ADR:8,II
:GHS05
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3700 to 1.3730 (20°C, 589 nm)
- Water:≤0.04 %
- Residue after evaporation:≤0.01 %
- Titration with NaOH:≥99.0 %