β-(N-Morpholino)ethanesulphonic acid (MES), (max. 1% H₂O) 99+%
:4432-31-9
:
:C₆H₁₃NO₄S
:MFCD00006181
:195.24 g/mol
:>300 °C (dec.)
:Ambient
- Appearance:White Crystals
- Infrared spectrum:Conforms
- Water:≤1.0 % (K.F.)
- Solubility:(0.5 M in water) Clear colorless
- Titration with NaOH:≥99.0 % (On anhydrous basis)