Order Entry
Germany
ContactUsLinkComponent
Acetone-[D₆] 99.8 atom % D, AcroSeal® for NMR spectroscopy
Irritant
Flammable
Acetone-[D₆] 99.8 atom % D, AcroSeal® for NMR spectroscopy
Catalog # 217371000.
CAS Number:  
undefined
Irritant
Flammable
Acetone-[D₆] 99.8 atom % D, AcroSeal® for NMR spectroscopy
Catalog # 217371000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.03 % (Coulometric)
  • D-Enrichment:
    ≥99.75 %

Specification Test Results

About this item

 :
 :64.03 g/mol
Boiling Pt:55 °C (760 mmHg)
Melting Pt:–93 °C
Density:0.87 g/cm³
Flash Pt:–20 °C
UN:1090
ADR:3,II
 :GHS07
 :GHS02
Storage Temperature:Ambient
CAS Number:666-52-4
Formula:D₃CC(O)CD₃
MDL Number:MFCD00044635