Order Entry
Germany
ContactUsLinkComponent
Cobalt(II) acetate tetrahydrate 98-102% ACS
Health
Environment
Cobalt(II) acetate tetrahydrate 98-102% ACS
Catalog # ACRO197101000
CAS Number:  
undefined
Health
Environment
Cobalt(II) acetate tetrahydrate 98-102% ACS
Catalog # ACRO197101000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Appearance:
    Pink to red Crystalline powder or granules
  • Titration Complexometric:
    98.0 to 102.0 %
  • Chloride (Cl):
    ≤0.002 %
  • Nitrate (NO3):
    ≤0.01 %
  • Sulfate (SO4):
    ≤0.005 %
  • Calcium (Ca):
    ≤0.005 %
  • Copper (Cu):
    ≤0.002 %
  • Iron (Fe):
    ≤0.001 %
  • Lead (Pb):
    ≤0.001 %
  • Magnesium (Mg):
    ≤0.005 %
  • Nickel (Ni):
    ≤0.1 %
  • Potassium (K):
    ≤0.01 %
  • Sodium (Na):
    ≤0.05 %
  • Zinc (Zn):
    ≤0.01 %
  • Insoluble matter:
    ≤0.01 %

Specification Test Results

About this item

 :
 :249.08 g/mol
Melting Pt:140 °C
Density:1,7 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3077
ADR:9,III
 :GHS08
 :GHS09
CAS Number:6147-53-1
Formula:(H₃CCOO)₂Co·4H₂O
MDL Number:MFCD00149569