Order Entry
Germany
ContactUsLinkComponent
Platinum 99.999% (trace metals basis), powder
Flammable
Platinum 99.999% (trace metals basis), powder
Catalog # ACRO193710010
CAS Number:  
undefined
Flammable
Platinum 99.999% (trace metals basis), powder
Catalog # ACRO193710010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Grey Powder
  • Total trace metal impurities:
    ≤10 ppm

Specification Test Results

About this item

 :
 :195.08 g/mol
Boiling Pt:3827 °C (760 mmHg)
Melting Pt:1772 °C
Density:21,4 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3089
ADR:4.1,III
 :GHS02
CAS Number:7440-06-4
Formula:Pt
MDL Number:MFCD00011179