Order Entry
Germany
ContactUsLinkComponent
Borane dimethyl sulphide complex 2 M in toluene, AcroSeal®
Flammable
Corrosive
Irritant
Health
Borane dimethyl sulphide complex 2 M in toluene, AcroSeal®
Certificates
undefined
Flammable
Corrosive
Irritant
Health
Borane dimethyl sulphide complex 2 M in toluene, AcroSeal®

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Solution
  • Strength:
    1.90 to 2.20 M
  • Melting point:
    161 to 165 °C
  • Loss on drying:
    ≤0.3 % (105 °C, 2 h)
  • HPLC:
    ≥97.5 %

Specification Test Results