Order Entry
Germany
ContactUsLinkComponent
Polyvinyl alcohol, M.W. ∼ 146,000-186,000 98.0-98.8 %, hydrolysed
Polyvinyl alcohol, M.W. ∼ 146,000-186,000 98.0-98.8 %, hydrolysed
Certificates
undefined
Polyvinyl alcohol, M.W. ∼ 146,000-186,000 98.0-98.8 %, hydrolysed

Specification Test Results

  • Appearance:
    White to cream Granular powder
  • Infrared spectrum:
    Conforms
  • Viscosity:
    62 to 72 cP (4 % in water)
  • Degree of hydrolysis:
    98.0 to 98.8 mol %
  • Ash:
    ≤1.2 % (as Na2O)
  • pH:
    5.0 to 7.0 (4 % in water)
  • Volatile matter:
    ≤5 %

Specification Test Results

Related Information