Order Entry
Germany
ContactUsLinkComponent
Polyvinyl alcohol, M.W. ∼ 31,000-50,000 98.0-98.8 %, hydrolysed
Polyvinyl alcohol, M.W. ∼ 31,000-50,000 98.0-98.8 %, hydrolysed
Certificates
undefined
Polyvinyl alcohol, M.W. ∼ 31,000-50,000 98.0-98.8 %, hydrolysed
Certificates

Specification Test Results

  • Appearance:
    White to cream Granular powder
  • Infrared spectrum:
    Conforms
  • Viscosity:
    5.5 to 6.6 cP (4 % in water)
  • Degree of hydrolysis:
    98.0 to 98.8 mol %
  • Ash:
    ≤1.2 % (as Na2O)
  • pH:
    5.0 to 7.0 (4 % in water)
  • Volatile matter:
    ≤5 %

Specification Test Results

Related Information