
2-(2-(2-Chloroethoxy)ethoxy)ethanol 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5197-62-6
:
Formula:C₆H₁₃ClO₃
MDL Number:MFCD00002874
Boiling Pt:256…258 °C (760 mmHg)
Density:1.163 g/cm³
Flash Pt:107 °C
:168.62 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear light yellow Liquid
- Infrared spectrum:Conforms
- GC:≥94 %
- HPLC:≥97.5 %
- Specific optical rotation:+158° to +168° (20°C, 589 nm) (c=1, water) On dry substance