Order Entry
Germany
ContactUsLinkComponent
 
 
2,1,3-Benzothiadiazole 98%
 
undefined
 
2,1,3-Benzothiadiazole 98%

 

  • Appearance:
    Colorless to white to light yellow Low melting solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    42 to 47 °C
  • GC:
    ≥97.5 %