Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dihydro-1H-inden-2-amine hydrochloride
2,3-Dihydro-1H-inden-2-amine hydrochloride 98%
2,3-Dihydro-1H-inden-2-amine hydrochloride 98%
:
Thermo Fisher Scientific
2,3-Dihydro-1H-inden-2-amine hydrochloride 98%
:
Thermo Fisher Scientific
:
2338-18-3
:
:
C₉H₁₂ClN
:
MFCD00012549
:
169.65 g/mol
:
246…247 °C (dec.)
:
GHS07
:
Ambient
Appearance:
White to off-white Powder
Infrared spectrum:
Conforms
HPLC:
≥97.5 %
GC:
≥90 %
ACRO10386