Order Entry
Germany
ContactUsLinkComponent
Anti-F13A1 Rabbit Monoclonal Antibody [clone: EP3372]
Anti-F13A1 Rabbit Monoclonal Antibody [clone: EP3372]
Supplier:  IMMUNOLOGIC
Certificates
Anti-F13A1 Rabbit Monoclonal Antibody [clone: EP3372]
Supplier:  IMMUNOLOGIC

Specifications

  • Catalog No:
  • ILM7834-C1
  • ILM7834-C05
  • ILM7834-C01
  • Antigen symbol:
  • F13A1
  • F13A1
  • F13A1
  • Antigen name:
  • Coagulation factor XIII A chain
  • Coagulation factor XIII A chain
  • Coagulation factor XIII A chain
  • Antigen synonyms:
  • F13A
  • F13A
  • F13A
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Clone:
  • EP3372
  • EP3372
  • EP3372
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Rabbit
  • Gene ID:
  • 2162
  • 2162
  • 2162
  • Isotype:
  • IgG
  • IgG
  • IgG
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • Form:
  • Concentrated
  • Concentrated
  • Concentrated
  • Storage temperature:
  • 2°C - 8°C
  • 2°C - 8°C
  • 2°C - 8°C

Specifications

Related Information

Frequently Bought Together