ESD safety shoes, lace-up, Fun, Maverick Blue Low
: ELTEN
Sporty, lightweight, leather-free ESD safety shoe made from microfibre and mesh inserts, a padded tongue plus a slip resistant, dual-density PU/PU sole (Fun).
- Climate-regulating, breathable textile lining
- Replaceable insole; orthopaedic insoles possible
- Full length inlay sole (ESD Pro), black
Ideal for use in dry work areas such as industry, storage, transport, or assembly and where there is a risk of penetration from pointed and sharp objects.
Colour: Blue/white
: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC
- Catalog No:
- ELTN723351/45
- ELTN723351/44
- ELTN723351/43
- ELTN723351/42
- ELTN723351/41
- ELTN723351/40
- ELTN723351/48
- ELTN723351/39
- ELTN723351/38
- ELTN723351/47
- ELTN723351/46
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001