About this item
HyQ® CDM4CHO™ is a chemically-defined protein-free medium containing no animal derived components. It is designed to increase the process yields for the industrial manufacture of recombinant proteins using a variety of CHO cell clones. HyQ® CDM4CHO™ has been successfully tested in various culture systems, including T-flasks, shaker flasks and bioreactors. HyQ® CDM4CHO™ contains Pluronic® F68 and L-glutamine, and does not contain phenol red.
- Metabolically designed for high cell yield and recombinant protein production
- Allows for direct or sequential adaptation
- Designed for large-scale culture applications, including perfusion and fed-batch strategies
- All components fully traceable
- Manufactured under cGMP
HyClone CDM4CHO contains Pluronic F68 and L-glutamine, and does not contain phenol red. It is also available without L-glutamine to support the GS gene expression system.