HyClone™ CDM4MAb Cell Culture Media
Supplier: HyClone products (Cytiva)
Certificates
About this item
ADCF medium for use with hybridoma and myeloma cell lines.
- Animal-derived component-free.
- Chemically defined formulation
- Designed for high cell yield and monoclonal antibody (MAb) production.
- Allows for direct or sequential adaptation
- Manufactured from traceable components according to cGMP guidelines
HyClone CDM4MAb is a chemically defined and animal-derived component-free (ADCF) medium. This regulatory-friendly medium is designed to increase process yields for the manufacture of antibodies and antibody fragments for therapeutic use in a variety of engineered hybridoma and myeloma cell lines.
CDM4MAb medium has been successfully tested in a variety of culture systems, including T-flasks, shaker flasks, and bioreactors including fed-batch and perfusion cultures. CDM4MAb medium does not contain phenol red and is available with or without L-glutamine and poloxamer 188. CDM4MAb is available in liquid and powder formats in user-friendly packaging.