Orders LinkContactUsLinkComponent
Fixable viability staining kits
Fixable viability staining kits
Supplier:   Biotium
Fixable viability staining kits
Supplier:  Biotium
Certificates
Fixable viability staining kits
Fixable viability staining kits
17 Options Available Below
Product Details & Documents
Live-or-Dye™ Fixable Viability Staining Kits are designed for discrimination between live and dead cells during flow cytometry or microscopy.
  • Highly dead-cell specific
  • Covalent labelling with reactive dyes
  • Withstand fixation and permeabilisation
  • The most available colours
  • Dye options designed for spectral flow
Live-or-Dye™ Fixable Viability Stains are cell membrane impermeable amine-reactive dyes. The dyes are able to enter into dead cells that have compromised membrane integrity and covalently label free amines on intracellular proteins. On live cells, the dyes react with surface proteins, but these are much less abundant than intracellular proteins, resulting in low staining levels compared to dead cells. Live-or-Dye™ labelling is extremely stable, allowing the cells to be fixed and permeabilised without loss of fluorescence or dye transfer between cells. The Live-or-Dye™ staining protocol has been optimised to maximise live/dead discrimination with minimal live cell staining, in order to prevent interference with immunostaining.
Live-or-Dye™ Fixable Viability Stains are cell membrane impermeable amine-reactive dyes. The dyes are able to enter into dead cells that have compromised membrane integrity and covalently label free amines on intracellular proteins. On live cells, the dyes react with surface proteins, but these are much less abundant than intracellular proteins, resulting in low staining levels compared to dead cells. Live-or-Dye™ labelling is extremely stable, allowing the cells to be fixed and permeabilised without loss of fluorescence or dye transfer between cells. The Live-or-Dye™ staining protocol has been optimised to maximise live/dead discrimination with minimal live cell staining, in order to prevent interference with immunostaining.
The dyes are named for their excitation/emission wavelengths. Choose from a wide variety of colours with fluorescence from UV to near-IR. Live-or-Dye™ 488/515, 568/583, 594/614, & 640/662 can be used for either flow cytometry or fluorescence microscopy. Live-or-Dye™ 510/550 & 665/685 have been validated for use in spectral flow cytometry on the Cytek® Aurora. Live-or-Dye™ 787/808 offers the best performance for the 808 laser line, effectively freeing up the channels in the visible spectrum.
Kit Components:
Live-or-Dye™ Fixable Viability Dye
Anhydrous DMSO
Ordering information: Number of assays is based on 1 ml reaction size (106cells); actual number of assays may vary depending on staining volume.
Product Family Options
Product Information
  • Laser line
    Pk
    Availability
    Price
  • BTIU32002-T
    Live-or-Dye™ 350/448, DAPI or violet emission filter, for flow cytometry, (λExEm) 347/448 nm
    Live-or-Dye™ 350/448, DAPI or violet emission filter, for flow cytometry, (λExEm) 347/448 nm
    355 nm
    50 Tests
    Log in for availability
    Log in for availability
    1 * 50 Tests
    1 * 50 Tests - 116,00 €
    116,00 €
    1 * 50 Tests
       116,00 €
  • BTIU32002
    Live-or-Dye™ 350/448, DAPI or violet emission filter, for flow cytometry, (λExEm) 347/448 nm
    Live-or-Dye™ 350/448, DAPI or violet emission filter, for flow cytometry, (λExEm) 347/448 nm
    355 nm
    200 Tests
    Log in for availability
    Log in for availability
    1 * 200 Tests
    1 * 200 Tests - 385,00 €
    385,00 €
    1 * 200 Tests
       385,00 €
  • BTIU32003-T
    Live-or-Dye™ 405/452, pacific blue emission filter, for flow cytometry, (λExEm) 408/452 nm
    Live-or-Dye™ 405/452, pacific blue emission filter, for flow cytometry, (λExEm) 408/452 nm
    405 nm
    50 Tests
    Log in for availability
    Log in for availability
    1 * 50 Tests
    1 * 50 Tests - 116,00 €
    116,00 €
    1 * 50 Tests
       116,00 €
  • BTIU32003
    Live-or-Dye™ 405/452, pacific blue emission filter, for flow cytometry, (λExEm) 408/452 nm
    Live-or-Dye™ 405/452, pacific blue emission filter, for flow cytometry, (λExEm) 408/452 nm
    405 nm
    200 Tests
    Log in for availability
    Log in for availability
    1 * 200 Tests
    1 * 200 Tests - 385,00 €
    385,00 €
    1 * 200 Tests
       385,00 €
  • BTIU32004-T
    Live-or-Dye™ 488/515, FITC emission filter, for flow cytometry and microscopy, (λExEm) 490/515 nm
    Live-or-Dye™ 488/515, FITC emission filter, for flow cytometry and microscopy, (λExEm) 490/515 nm
    488 nm
    50 Tests
    Log in for availability
    Log in for availability
    1 * 50 Tests
    1 * 50 Tests - 116,00 €
    116,00 €
    1 * 50 Tests
       116,00 €
  • BTIU32004
    Live-or-Dye™ 488/515, FITC emission filter, for flow cytometry, (λExEm) 490/515 nm
    Live-or-Dye™ 488/515, FITC emission filter, for flow cytometry, (λExEm) 490/515 nm
    488 nm
    200 Tests
    Log in for availability
    Log in for availability
    1 * 200 Tests
    1 * 200 Tests - 385,00 €
    385,00 €
    1 * 200 Tests
       385,00 €
  • BTIU32005-T
    Live-or-Dye™ 568/583, PE emission filter, for flow cytometry and microscopy, (λExEm) 562/583 nm
    Live-or-Dye™ 568/583, PE emission filter, for flow cytometry and microscopy, (λExEm) 562/583 nm
    488 or 561 nm
    50 Tests
    Log in for availability
    Log in for availability
    1 * 50 Tests
    1 * 50 Tests - 116,00 €
    116,00 €
    1 * 50 Tests
       116,00 €
  • BTIU32005
    Live-or-Dye™ 568/583, PE emission filter, for flow cytometry and microscopy, (λExEm) 562/583 nm
    Live-or-Dye™ 568/583, PE emission filter, for flow cytometry and microscopy, (λExEm) 562/583 nm
    488 or 561 nm
    200 Tests
    Log in for availability
    Log in for availability
    1 * 200 Tests
    1 * 200 Tests - 385,00 €
    385,00 €
    1 * 200 Tests
       385,00 €
  • BTIU32006-T
    Live-or-Dye™ 594/614, PE-Texas red emission filter, for flow cytometry and microscopy, (λExEm) 561/624 nm
    Live-or-Dye™ 594/614, PE-Texas red emission filter, for flow cytometry and microscopy, (λExEm) 561/624 nm
    488 or 561 nm
    50 Tests
    Log in for availability
    Log in for availability
    1 * 50 Tests
    1 * 50 Tests - 116,00 €
    116,00 €
    1 * 50 Tests
       116,00 €
  • BTIU32006
    Live-or-Dye™ 594/614, PE-Texas red emission filter, for flow cytometry and microscopy, (λExEm) 561/624 nm
    Live-or-Dye™ 594/614, PE-Texas red emission filter, for flow cytometry and microscopy, (λExEm) 561/624 nm
    488 or 561 nm
    200 Tests
    Log in for availability
    Log in for availability
    1 * 200 Tests
    1 * 200 Tests - 385,00 €
    385,00 €
    1 * 200 Tests
       385,00 €
  • BTIU32007-T
    Live-or-Dye™ 640/662, APC emission filter, for flow cytometry and microscopy, (λExEm) 642/662 nm
    Live-or-Dye™ 640/662, APC emission filter, for flow cytometry and microscopy, (λExEm) 642/662 nm
    633 or 640 nm
    50 Tests
    Log in for availability
    Log in for availability
    1 * 50 Tests
    1 * 50 Tests - 116,00 €
    116,00 €
    1 * 50 Tests
       116,00 €
  • BTIU32007
    Live-or-Dye™ 640/662, APC emission filter, for flow cytometry and microscopy, (λExEm) 642/662 nm
    Live-or-Dye™ 640/662, APC emission filter, for flow cytometry and microscopy, (λExEm) 642/662 nm
    633 or 640 nm
    200 Tests
    Log in for availability
    Log in for availability
    1 * 200 Tests
    1 * 200 Tests - 385,00 €
    385,00 €
    1 * 200 Tests
       385,00 €
  • BTIU32008-T
    Live-or-Dye™ 750/777, APC-Cy7 emission filter, for flow cytometry, (λExEm) 755/777 nm
    Live-or-Dye™ 750/777, APC-Cy7 emission filter, for flow cytometry, (λExEm) 755/777 nm
    633 or 640 nm
    50 Tests
    Log in for availability
    Log in for availability
    1 * 50 Tests
    1 * 50 Tests - 116,00 €
    116,00 €
    1 * 50 Tests
       116,00 €
  • BTIU32008
    Live-or-Dye™ 750/777, APC-Cy7 emission filter, for flow cytometry, (λExEm) 755/777 nm
    Live-or-Dye™ 750/777, APC-Cy7 emission filter, for flow cytometry, (λExEm) 755/777 nm
    633 or 640 nm
    200 Tests
    Log in for availability
    Log in for availability
    1 * 200 Tests
    1 * 200 Tests - 385,00 €
    385,00 €
    1 * 200 Tests
       385,00 €
  • BTIU32009-T
    Live-or-Dye™ 405/545, AmCyan emission filter, for flow cytometry, (λExEm) 395/545 nm
    Live-or-Dye™ 405/545, AmCyan emission filter, for flow cytometry, (λExEm) 395/545 nm
    405 nm
    50 Tests
    Log in for availability
    Log in for availability
    1 * 50 Tests
    1 * 50 Tests - 116,00 €
    116,00 €
    1 * 50 Tests
       116,00 €
  • BTIU32009
    Live-or-Dye™ 405/545, AmCyan emission filter, for flow cytometry, (λExEm) 395/545 nm
    Live-or-Dye™ 405/545, AmCyan emission filter, for flow cytometry, (λExEm) 395/545 nm
    405 nm
    200 Tests
    Log in for availability
    Log in for availability
    1 * 200 Tests
    1 * 200 Tests - 385,00 €
    385,00 €
    1 * 200 Tests
       385,00 €
  • 32012.
    Live-or-Dye™ 510/550, for spectral cytometry
    Live-or-Dye™ 510/550, for spectral cytometry
    488 or 532 nm
    200 Tests
    Log in for availability
    Log in for availability
    1 * 200 Tests
    1 * 200 Tests - 385,00 €
    385,00 €
    1 * 200 Tests
       385,00 €