Dimethyl carbonate, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:616-38-6
:
Formula:C₃H₆O₃
MDL Number:MFCD00008420
:90.08 g/mol
Boiling Pt:90 °C (1013 hPa)
Melting Pt:2…4 °C
Density:1.069 g/cm³
Flash Pt:18 °C
Storage Temperature:Ambient
UN:1161
ADR:3,II
:GHS02
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥99.0 %
- Color scale:≤30 APHA