
2-Bromo-6-methoxypyridine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:40473-07-2
:
Formula:C₆H₆BrNO
MDL Number:MFCD00088345
:188.02 g/mol
Boiling Pt:206 °C
Flash Pt:104 °C
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Clear colorless to golden Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %