2,6-Dimethylbenzaldehyde 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1123-56-4
:
Formula:C₉H₁₀O
MDL Number:MFCD00128003
:134.18 g/mol
Melting Pt:27…30 °C
Flash Pt:96 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to yellow Liquid or low melting solid
- Infrared spectrum:Conforms
- GC:≥96.0 %