

3-Hydroxythiophenol 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:40248-84-8
:
Formula:C₆H₆OS
MDL Number:MFCD01317829
:126.18 g/mol
Boiling Pt:135 °C (35 mmHg)
Melting Pt:16.5 °C
Density:1.63 g/cm³
Storage Temperature:Ambient
UN:2922
ADR:8,II
:GHS07
:GHS05
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- GC:≥95 %