

(R)-(+)-1-Boc-3-aminopyrrolidine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:147081-49-0
:
Formula:C₉H₁₈N₂O₂
MDL Number:MFCD03419272
:186.25 g/mol
Flash Pt:91 °C
UN:2810
ADR:6.1,III
:GHS06
:GHS05
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear light yellow Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
- Specific optical rotation:+2° to +4° (20°C, 589 nm) (c=1 in CHCl3)